Learn More
Teknova Sabouraud Dextrose Agar Plates with Chloramphenicol-50. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388186
Encompass
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.
Catalog No. 50-238-8186 Supplier Teknova Supplier No. S2043
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.