Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Sabouraud Dextrose Agar (SabDex)
SDP

Catalog No. 23001775 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-775 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-775 Supplier Hardy Diagnostics™ Supplier No. W250
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sabouraud Dextrose Agar is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts

  • The pH is adjusted to approximately 5.6 in order to enhance the growth of fungi, especially dermatophytes, and to slightly inhibit bacterial growth in clinical specimens
  • This medium is recommended for mold and yeast counts by the Association of Official Analytical Chemists
  • Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts

Specifications

Appearance Translucent
Color Light Amber
Detects Fungi, Yeast, Mold
Format Plate
Product Type Agar Plate
Target Organism Class Fungi
Test Type Microbial Cultivation
Fill Volume 60 mL
For Use With (Application) Isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts
Incubation Conditions 20°C to 25°C, Aerobic
Incubation Time 1 to 5 days
No. per Pack 5
Packaging Type Pack
Quantity 5 Ea.
Dimensions (Dia. x H) 100 x 25 mm
Height (Metric) 25 mm
Diameter (Metric) 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.