Learn More
Alkali Scientific Sabouraud Dextrose Agar (SDA) Emmons w/ 0.1% Chloramphenicol & 1.5% Tween 80 Bottle, 10/Pk
SDP
Catalog No. 5000662827
Encompass_Preferred
Sabouraud Dextrose Agar with Tween 80 and Chloramphenicol is a specialized culture medium designed for isolating lipid dependent fungi from clinical or environmental samples. The formulation incorporates Tween 80, which provides fatty acids required by certain fungi that cannot synthe their own lipids. This makes the medium particularly useful for cultivating organisms such as Malassezia species. Chloramphenicol is included as a broad spectrum antibiotic to suppress bacterial growth that might otherwise interfere with fungal detection. The combination of lipid supplementation and antibacterial protection creates conditions that allow slow growing or fastidious fungi to develop successfully during incubation.
Catalog No. 50-006-62827 Supplier Alkali Scientific Supplier No. SDAT02
Sabouraud Dextrose Agar with Tween 80 and Chloramphenicol is a specialized culture medium designed for isolating lipid dependent fungi from clinical or environmental samples. The formulation incorporates Tween 80, which provides fatty acids required by certain fungi that cannot synthe their own lipids. This makes the medium particularly useful for cultivating organisms such as Malassezia species. Chloramphenicol is included as a broad spectrum antibiotic to suppress bacterial growth that might otherwise interfere with fungal detection. The combination of lipid supplementation and antibacterial protection creates conditions that allow slow growing or fastidious fungi to develop successfully during incubation.