Learn More
Alkali Scientific Sabouraud Dextrose Agar (SDA) Slant 14mL Tube, 10/Pk
SDP
Catalog No. 5000663115
Encompass_Preferred
Sabouraud Dextrose Agar is a standard culture medium used for the isolation and cultivation of yeasts, molds, and dermatophytes. This 14mL slant format provides a larger agar volume and increased surface area compared to traditional slants. The deeper agar reservoir helps maintain moisture during extended incubation periods, which are often required for slow growing fungal species. The medium contains a high concentration of dextrose and a slightly acidic pH that favors fungal growth while limiting bacterial contamination. The larger slant surface also allows clearer observation of colony characteristics such as pigment production, aerial mycelium, and colony texture, which are important for fungal identification and long term culture maintenance.
Catalog No. 50-006-63115 Supplier Alkali Scientific Supplier No. SDAT01
Sabouraud Dextrose Agar is a standard culture medium used for the isolation and cultivation of yeasts, molds, and dermatophytes. This 14mL slant format provides a larger agar volume and increased surface area compared to traditional slants. The deeper agar reservoir helps maintain moisture during extended incubation periods, which are often required for slow growing fungal species. The medium contains a high concentration of dextrose and a slightly acidic pH that favors fungal growth while limiting bacterial contamination. The larger slant surface also allows clearer observation of colony characteristics such as pigment production, aerial mycelium, and colony texture, which are important for fungal identification and long term culture maintenance.