Learn More
Alkali Scientific Sabouraud Dextrose Agar (SDA) w/ 5% Soy Lecithin & 4% Tween 20 100mL Bottle, 12/CS
SDP
Catalog No. 5000662718
Encompass_Preferred
Sabouraud Dextrose Agar with Soy Lecithin and Tween 20 is a specialized neutralizing medium designed for environmental monitoring in pharmaceutical and industrial facilities. The formulation contains powerful neutralizers that deactivate residues from disinfectants and cleaning chemicals commonly used on laboratory and manufacturing surfaces. Soy lecithin helps neutralize quaternary ammonium compounds, while Tween 20 neutralizes phenolic disinfectants and related chemicals. These additives prevent residual disinfectants from killing microorganisms during testing, ensuring accurate recovery of fungi from sampled surfaces. The medium is widely used in environmental monitoring programs where reliable detection of microbial contamination is essential.
Catalog No. 50-006-62718 Supplier Alkali Scientific Supplier No. SDAB02
Sabouraud Dextrose Agar with Soy Lecithin and Tween 20 is a specialized neutralizing medium designed for environmental monitoring in pharmaceutical and industrial facilities. The formulation contains powerful neutralizers that deactivate residues from disinfectants and cleaning chemicals commonly used on laboratory and manufacturing surfaces. Soy lecithin helps neutralize quaternary ammonium compounds, while Tween 20 neutralizes phenolic disinfectants and related chemicals. These additives prevent residual disinfectants from killing microorganisms during testing, ensuring accurate recovery of fungi from sampled surfaces. The medium is widely used in environmental monitoring programs where reliable detection of microbial contamination is essential.