Learn More
Alkali Scientific Sabouraud Dextrose Agar (SDA) w/ Lecithin & Polysorbate 80 (Tween 80), 14mL Contact Plates, 5 Day QC, 10/Pk
SDP
Catalog No. 5000663118
Encompass_Preferred
Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 is a specialized mycological medium designed for environmental surface monitoring in cleanrooms and other controlled environments. The 14 mL contact plate format features a convex agar surface that enables direct sampling of flat surfaces for the detection of fungal and yeast contamination. Lecithin and Polysorbate 80 neutralize common disinfectant residues, including quaternary ammonium compounds and phenolics, improving the recovery of microorganisms from sanitized surfaces. This medium supports the growth of fungi, yeasts, and molds commonly encountered in pharmaceutical manufacturing and healthcare facilities. Plates are terminally sterilized, quality tested, and released with validated sterility and performance testing to ensure reliable environmental monitoring results.
Catalog No. 50-006-63118 Supplier Alkali Scientific Supplier No. SDAP00
Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 is a specialized mycological medium designed for environmental surface monitoring in cleanrooms and other controlled environments. The 14 mL contact plate format features a convex agar surface that enables direct sampling of flat surfaces for the detection of fungal and yeast contamination. Lecithin and Polysorbate 80 neutralize common disinfectant residues, including quaternary ammonium compounds and phenolics, improving the recovery of microorganisms from sanitized surfaces. This medium supports the growth of fungi, yeasts, and molds commonly encountered in pharmaceutical manufacturing and healthcare facilities. Plates are terminally sterilized, quality tested, and released with validated sterility and performance testing to ensure reliable environmental monitoring results.