Learn More
Alkali Scientific Sabouraud Dextrose Agar (SDA) W/Thiamphenicol & Oxytetracyline 20mL Plate, 10/Pk
SDP
Catalog No. 5000662764
Encompass_Preferred
Sabouraud Dextrose Agar with Thiamphenicol and Oxytetracycline is a highly selective culture medium developed for isolating fungi from samples heavily contaminated with bacteria. The formulation combines two broad spectrum antibiotics that inhibit bacterial protein synthesis and suppress both Gram positive and Gram negative bacteria. This dual antibiotic system provides stronger bacterial inhibition than standard Sabouraud agar, allowing slow growing fungal colonies to develop without interference. The medium is commonly used in clinical diagnostics, environmental testing, and microbiological analysis where fungal pathogens must be recovered from complex mixed microbial populations.
Catalog No. 50-006-62764 Supplier Alkali Scientific Supplier No. SDAP03
Sabouraud Dextrose Agar with Thiamphenicol and Oxytetracycline is a highly selective culture medium developed for isolating fungi from samples heavily contaminated with bacteria. The formulation combines two broad spectrum antibiotics that inhibit bacterial protein synthesis and suppress both Gram positive and Gram negative bacteria. This dual antibiotic system provides stronger bacterial inhibition than standard Sabouraud agar, allowing slow growing fungal colonies to develop without interference. The medium is commonly used in clinical diagnostics, environmental testing, and microbiological analysis where fungal pathogens must be recovered from complex mixed microbial populations.