Learn More
Alkali Scientific Sabouraud Dextrose Agar w/Chloramphenicol 20mL Plate, 10/Pk
SDP
Catalog No. 5000662739
Encompass_Preferred
Sabouraud Dextrose Agar with Chloramphenicol is a selective fungal culture medium used for the isolation of yeasts and molds from samples that may contain large numbers of bacteria. While the acidic pH of standard Sabouraud agar already discourages many bacterial species, the addition of chloramphenicol provides an extra level of selectivity. This broad spectrum antibiotic inhibits most Gram positive and Gram negative bacteria, preventing them from overgrowing slower developing fungal colonies. The 20mL pre poured plate format offers a convenient ready to use solution that eliminates media preparation steps. It is widely used in clinical laboratories, environmental monitoring, and microbiological testing of contaminated specimens.
Catalog No. 50-006-62739 Supplier Alkali Scientific Supplier No. SDSP00
Sabouraud Dextrose Agar with Chloramphenicol is a selective fungal culture medium used for the isolation of yeasts and molds from samples that may contain large numbers of bacteria. While the acidic pH of standard Sabouraud agar already discourages many bacterial species, the addition of chloramphenicol provides an extra level of selectivity. This broad spectrum antibiotic inhibits most Gram positive and Gram negative bacteria, preventing them from overgrowing slower developing fungal colonies. The 20mL pre poured plate format offers a convenient ready to use solution that eliminates media preparation steps. It is widely used in clinical laboratories, environmental monitoring, and microbiological testing of contaminated specimens.