Learn More
Alkali Scientific Sabouraud Dextrose Agar w/Lecithin & Tween 80 14mL Contact Locking Lid Plate, 10/Pk
SDP
Catalog No. 5000662712
Encompass_Preferred
Sabouraud Dextrose Agar with Lecithin and Tween 80 is a specialized contact plate designed for environmental monitoring of surfaces in pharmaceutical, clinical, and food production facilities. The plate contains a slightly raised agar surface that allows technicians to press the medium directly against surfaces to collect microorganisms for testing. Lecithin and Tween 80 act as neutralizing agents that inactivate residues from disinfectants and cleaning chemicals. This ensures that microorganisms collected from sanitized surfaces remain viable during incubation. The plate design often includes a grid pattern that helps laboratories count colonies and estimate microbial contamination levels on tested surfaces.
Catalog No. 50-006-62712 Supplier Alkali Scientific Supplier No. SDSP01
Sabouraud Dextrose Agar with Lecithin and Tween 80 is a specialized contact plate designed for environmental monitoring of surfaces in pharmaceutical, clinical, and food production facilities. The plate contains a slightly raised agar surface that allows technicians to press the medium directly against surfaces to collect microorganisms for testing. Lecithin and Tween 80 act as neutralizing agents that inactivate residues from disinfectants and cleaning chemicals. This ensures that microorganisms collected from sanitized surfaces remain viable during incubation. The plate design often includes a grid pattern that helps laboratories count colonies and estimate microbial contamination levels on tested surfaces.