Learn More
Alkali Scientific Sabouraud Dextrose Broth, 500 Grams
SDP
Catalog No. 5000659996
Encompass_Preferred
Sabouraud Dextrose Broth is a liquid medium designed for the cultivation of yeasts, molds, and other fungi. The 500g package provides a sufficient volume for preparing multiple batches of broth, ideal for large-scale fungal culture in clinical, environmental, and food industry applications. The broth contains dextrose to support fungal growth and is typically used in sterility testing, fungal pathogen isolation, and enrichment procedures. The neutral pH helps promote the growth of fungi while inhibiting most bacteria. Sabouraud Dextrose Broth is commonly used for the enrichment of fungal species before transfer to solid media for isolation and further identification.
Catalog No. 50-006-59996 Supplier Alkali Scientific Supplier No. OUR001CP
Sabouraud Dextrose Broth is a liquid medium designed for the cultivation of yeasts, molds, and other fungi. The 500g package provides a sufficient volume for preparing multiple batches of broth, ideal for large-scale fungal culture in clinical, environmental, and food industry applications. The broth contains dextrose to support fungal growth and is typically used in sterility testing, fungal pathogen isolation, and enrichment procedures. The neutral pH helps promote the growth of fungi while inhibiting most bacteria. Sabouraud Dextrose Broth is commonly used for the enrichment of fungal species before transfer to solid media for isolation and further identification.