Learn More
Teknova Sabouraud Dextrose Broth. 500mL, Sterile. 4 pack.
SDP
Catalog No. 502387523
Encompass
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.
Catalog No. 50-238-7523 Supplier Teknova Supplier No. S58054PK
Sabouraud dextrose agar is a selective medium used to isolate yeast and fungi while permitting the growth of filamentous bacteria like Nocardia.