Learn More
NEOGEN Sabouraud Dextrose Broth, 5 kg
SDP
Catalog No. 502015577
Encompass_Preferred
Sabouraud Dextrose Broth is used for the cultivation of fungi from sterile and non-sterile products. Sabouraud Dextrose Broth conforms to Harmonized USP/EP/JP Requirements and is not intended for use in the diagnosis of disease or other conditions in humans.A medium recommended by the Harmonized Pharmacopoeia for the enrichment of Candida albicans from non-sterile samples and fungal test strain cultivation. Conforms to USP/EP/JP performance specifications. The peptones and dextrose provide a nutritious base for luxuriant fungal growth and the acidic pH affords selectivity against bacteria. Due to the high carbohydrate content and low pH this medium is highly sensitive to overheating. According to the Harmonized Pharmacopoeia, Sabouraud Dextrose Broth is used as an enrichment broth, with subculture performed onto Sabouraud Dextrose Agar.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-broth
Catalog No. 50-201-5577 Supplier NEOGEN Supplier No. 700004536
Sabouraud Dextrose Broth is used for the cultivation of fungi from sterile and non-sterile products. Sabouraud Dextrose Broth conforms to Harmonized USP/EP/JP Requirements and is not intended for use in the diagnosis of disease or other conditions in humans.A medium recommended by the Harmonized Pharmacopoeia for the enrichment of Candida albicans from non-sterile samples and fungal test strain cultivation. Conforms to USP/EP/JP performance specifications. The peptones and dextrose provide a nutritious base for luxuriant fungal growth and the acidic pH affords selectivity against bacteria. Due to the high carbohydrate content and low pH this medium is highly sensitive to overheating. According to the Harmonized Pharmacopoeia, Sabouraud Dextrose Broth is used as an enrichment broth, with subculture performed onto Sabouraud Dextrose Agar.For more information visit https://www.neogen.com/solutions/Microbiology/sabouraud-dextrose-broth