Learn More
Alkali Scientific Sabouraud Dextrose Broth (SAB) 1000mL Bottle, 1/Pk
SDP
Catalog No. 5000662293
Encompass_Preferred
Sabouraud Dextrose Broth is a sterile liquid culture medium used for the enrichment and cultivation of yeasts, molds, and other acid tolerant microorganisms. It contains a high concentration of dextrose to provide an abundant energy source for fungal metabolism and growth. The broth is maintained at an acidic pH of approximately 5.6, which discourages many bacterial species while allowing fungi to develop more readily. This liquid formulation is widely used in sterility testing, environmental monitoring, and microbiological quality control. It is particularly useful when recovering fungal pathogens from contaminated samples or pharmaceutical products where low levels of fungi must be detected before further isolation or identification procedures.
Catalog No. 50-006-62293 Supplier Alkali Scientific Supplier No. B1325
Sabouraud Dextrose Broth is a sterile liquid culture medium used for the enrichment and cultivation of yeasts, molds, and other acid tolerant microorganisms. It contains a high concentration of dextrose to provide an abundant energy source for fungal metabolism and growth. The broth is maintained at an acidic pH of approximately 5.6, which discourages many bacterial species while allowing fungi to develop more readily. This liquid formulation is widely used in sterility testing, environmental monitoring, and microbiological quality control. It is particularly useful when recovering fungal pathogens from contaminated samples or pharmaceutical products where low levels of fungi must be detected before further isolation or identification procedures.