Learn More
Alkali Scientific Sabouraud Dextrose Broth (SAB) 100mL, 1/Pk
SDP
Supplier:  Alkali Scientific SABB01
Encompass_Preferred
Sabouraud Dextrose Broth is a sterile liquid medium formulated for the cultivation and enrichment of yeasts, molds, and other acid tolerant microorganisms. This 100mL bottle format provides a convenient single use volume that allows laboratories to inoculate samples directly into the medium without additional transfers. The broth contains a high concentration of dextrose that supports rapid fungal metabolism and growth. Its acidic pH helps suppress many bacterial contaminants, allowing fungi to recover and multiply more easily in mixed samples. This format is widely used in sterility testing, food microbiology, and pharmaceutical quality control where samples are added directly to the broth for incubation and detection of fungal contamination.
Catalog No. 50-006-63293