Learn More
Alkali Scientific Sabouraud Dextrose Broth (SAB) 9mL Tube, 10/Pk
SDP
Catalog No. 5000663343
Encompass_Preferred
Sabouraud Dextrose Broth is a sterile liquid medium commonly used for the cultivation, isolation, and maintenance of yeasts and molds in microbiology laboratories. This 9mL tube format combines fungal enrichment properties with a volume specifically designed for small scale dilution or culture preparation. The formulation contains a high concentration of dextrose to provide an abundant energy source that promotes fungal metabolism and growth. The acidic pH environment discourages many bacterial species while supporting fungal development. These tubes are commonly used for preparing dilutions of samples, reviving fungal cultures, and incubating small inoculated samples during routine laboratory analysis and microbial identification procedures.
Catalog No. 50-006-63343 Supplier Alkali Scientific Supplier No. SABT00
Sabouraud Dextrose Broth is a sterile liquid medium commonly used for the cultivation, isolation, and maintenance of yeasts and molds in microbiology laboratories. This 9mL tube format combines fungal enrichment properties with a volume specifically designed for small scale dilution or culture preparation. The formulation contains a high concentration of dextrose to provide an abundant energy source that promotes fungal metabolism and growth. The acidic pH environment discourages many bacterial species while supporting fungal development. These tubes are commonly used for preparing dilutions of samples, reviving fungal cultures, and incubating small inoculated samples during routine laboratory analysis and microbial identification procedures.