Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Sabouraud Dextrose Broth (SabDex)
SDP

Catalog No. 23001829 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-829 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-829 Supplier Hardy Diagnostics™ Supplier No. K54
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.

  • Sabouraud Dextrose Agar, Sabouraud Dextrose Broth, and Sabouraud Dextrose Agar, Emmons are recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.

Specifications

Broth Type Sabouraud Dextrose Broth
Manufacturing Origin United States
Product Type Broth Tube
Sterility Non-sterile
Target Organism Class Fungi, Yeast
Test Type Microbial Detection
Packaging Type Glass Tube
Quantity 10 mL
Dimensions (Dia. x H) 16 x 125 mm
Dimensions (L x W) 15 x 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.