Learn More
Research Products International Corp Sabouraud Dextrose Broth, 500 G
SDP
Catalog No. 502252951
Encompass_Preferred
Directions: Dissolve 30g in 1L distilled water. Heat and stir to completely dissolve agar. Sterilize by autoclaving for 15 minutes.
Formulation
Casein Digest Peptone: 10.0 g/L
Dextrose: 20.0 g/L
This item is not returnable. View return policy
Catalog No. 50-225-2951 Supplier Research Products International Corp Supplier No. S12520500.0
Directions: Dissolve 30g in 1L distilled water. Heat and stir to completely dissolve agar. Sterilize by autoclaving for 15 minutes.
Formulation
Casein Digest Peptone: 10.0 g/L
Dextrose: 20.0 g/L