Learn More
General Laboratory Products Sabouraud Dextrose Agar (SDA), Monoplates, 10/pk
Catalog No. NC0416820
Encompass_Preferred
Sabouraud Dextrose Agar - Monoplates, 100x15mm, Deep Fill, 10 per pack. Base for the Cultivation of Fungi and Aciduric Microorganisms.
Catalog No. NC0416820 Supplier General Laboratory Products Supplier No. MP3200DF
Sabouraud Dextrose Agar - Monoplates, 100x15mm, Deep Fill, 10 per pack. Base for the Cultivation of Fungi and Aciduric Microorganisms.