Learn More
Medchemexpress LLC Sabutoclax | 1228108-65-3 | C42H40N2O8 | 5 MG
SDP
Catalog No. 5000372146
Encompass_Preferred
Sabutoclax is a potent and effective inhibitor of the Bcl-2 Family, targeting Bcl-2, Bcl-XL, Mcl-1, and Bfl-1. It enhances the expression of pro-apoptotic proteins such as Bax, Bim, PUMA, and survivin, playing a role in inducing cell apoptosis and inhibiting cancer cell growth.
  • Potent and effective Bcl-2 family inhibitor.
  • Increases Bax, Bim, PUMA, and survivin expression.
  • Inhibits cell growth in human prostate and lung cancer cell lines.
  • Induces cell apoptosis in human diffuse large B-cell lymphoma cells.
  • Upregulates pro-apoptotic proteins in chemoresistant cells.
  • Reduces tumor growth in animal models.
Catalog No. 50-003-72146 Supplier Medchemexpress LLC Supplier No. HY151915MG
Sabutoclax is a potent and effective inhibitor of the Bcl-2 Family, targeting Bcl-2, Bcl-XL, Mcl-1, and Bfl-1. It enhances the expression of pro-apoptotic proteins such as Bax, Bim, PUMA, and survivin, playing a role in inducing cell apoptosis and inhibiting cancer cell growth.
  • Potent and effective Bcl-2 family inhibitor.
  • Increases Bax, Bim, PUMA, and survivin expression.
  • Inhibits cell growth in human prostate and lung cancer cell lines.
  • Induces cell apoptosis in human diffuse large B-cell lymphoma cells.
  • Upregulates pro-apoptotic proteins in chemoresistant cells.
  • Reduces tumor growth in animal models.