Learn More
Selleck Chemical LLC Sabutoclax 5mg 1228108-65-3 BI-97C1
SDP
Supplier:  Selleck Chemical LLC S80615MG
Encompass_Preferred
Sabutoclax (BI-97C1) is a pan-Bcl-2 inhibitor, including Bcl-xL, Bcl-2, Mcl-1 and Bfl-1 with IC50 of 0.31 ?M, 0.32 ?M, 0.20 ?M and 0.62 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0852869