Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saccharic acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 g/mol | C6H10O8 | 250 MG
SDP

Catalog No. 5000282075
Encompass_Preferred

Saccharic acid is a small-molecule inhibitor used in biochemical research to inhibit β-glucuronidase activity. It is supplied as a high-purity solid for enzymatic assays, cell-based studies, and other laboratory applications where selective inhibition of glucuronide hydrolysis is required.

  • Competitive inhibitor of β-glucuronidase for enzymatic and biochemical assays.
  • High purity: 98.0% suitable for research applications.
  • Chemical formula C6H10O8 and molecular weight 210.14 g/mol.
  • CAS number 87-73-0 for unambiguous identification.
  • Available as a 250 MG solid pack for laboratory use.
  • Stable solid suitable for standard storage and handling in research labs.

Catalog No. 50-002-82075 Supplier Medchemexpress LLC Supplier No. HYW345885250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saccharic acid is a small-molecule inhibitor used in biochemical research to inhibit β-glucuronidase activity. It is supplied as a high-purity solid for enzymatic assays, cell-based studies, and other laboratory applications where selective inhibition of glucuronide hydrolysis is required.

  • Competitive inhibitor of β-glucuronidase for enzymatic and biochemical assays.
  • High purity: 98.0% suitable for research applications.
  • Chemical formula C6H10O8 and molecular weight 210.14 g/mol.
  • CAS number 87-73-0 for unambiguous identification.
  • Available as a 250 MG solid pack for laboratory use.
  • Stable solid suitable for standard storage and handling in research labs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.