Learn More
Cayman Chemical Saccharin 1-methylimidazole, Purity: ≥98%, CAS: 482333-74-4, 5 mg
SDP
Catalog No. 502938688
Encompass_Preferred
A chemical activator that has been commonly used in solid-phase synthesis of DNA and RNA oligonucleotides; catalyzes the phosphoramidite coupling reaction of the 5' hydroxy of immobilized nucleosides and the amine group of nucleoside phosphoramidite; has been used to synthesize ssDNA oligonucleotides that are resistant to nuclease digestion
Catalog No. 50-293-8688 Supplier Cayman Chemical Supplier No. 292425 MG
A chemical activator that has been commonly used in solid-phase synthesis of DNA and RNA oligonucleotides; catalyzes the phosphoramidite coupling reaction of the 5' hydroxy of immobilized nucleosides and the amine group of nucleoside phosphoramidite; has been used to synthesize ssDNA oligonucleotides that are resistant to nuclease digestion