Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin Sodium, Dihydrate, Powder, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606120 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-120 500 g Poly Bottle
18-606-119 100 g Poly Bottle
18-606-121 2.5 kg Poly Pail
18-606-122 12 kg Poly Pail
18-606-124 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-120 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO200500GM
Add to Cart
Add to Cart

SO200, 6155-57-3, C7H4NNaO3S·2H2O

Spectrum™ Chemical Saccharin Sodium, Dihydrate, Powder, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6155-57-3
Molecular Formula C7H8NNaO5S
Molecular Weight (g/mol) 241.19
InChI Key NAUHOSQQKJTUAX-UHFFFAOYSA-M
IUPAC Name sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate
SMILES O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.