Learn More
Medchemexpress LLC HY-B1390B 500mg Medchemexpress, Saccharin (sodium hydrate) CAS:82385-42-0 Purity:>98%
SDP
Catalog No. 502082701
Encompass_Preferred
Medchemexpress, HY-B1390B 500mg Saccharin (sodium hydrate) CAS:82385-42-0 Saccharin sodium hydrate is an orally active, non-caloric artificial sweeteners (NAS). Saccharin sodium hydrate has bacteriostatic and microbiome-modulating properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2701 Supplier Medchemexpress LLC Supplier No. HYB1390B500MG
Medchemexpress, HY-B1390B 500mg Saccharin (sodium hydrate) CAS:82385-42-0 Saccharin sodium hydrate is an orally active, non-caloric artificial sweeteners (NAS). Saccharin sodium hydrate has bacteriostatic and microbiome-modulating properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.