Learn More
Cayman Chemical SaccharocarcIn A 2.5mg
SDP
Catalog No. 502939642
Encompass_Preferred
An antibiotic; active against M. luteus, S. aureus, E. coli, P. aeruginosa, and C. albicans in a disc assay; inhibits C. trachomatis infection in McCoy cells
Catalog No. 50-293-9642 Supplier Cayman Chemical Supplier No. 280942.5 MG
An antibiotic; active against M. luteus, S. aureus, E. coli, P. aeruginosa, and C. albicans in a disc assay; inhibits C. trachomatis infection in McCoy cells