Learn More
ATCC Hyphozyma roseonigra
Supplier:  ATCC 20624
Encompass
Hyphozyma roseonigra strain 30-1-1B is provided as an ampoule containing viable cells (may include spores and mycelia) suspended in cryoprotectant. This fungal type strain was isolated from soil in New Jersey and is known to produce diol and furan compounds.
Catalog No. 50-238-1659
 Restricted Item