Learn More
ATCC Saccharomyces cerevisiae strain EBY100 (ATCC MYA-4941™)
Catalog No. 502383727
Encompass
Saccharomyces cerevisiae strain EBY100 (ATCC MYA-4941™) is a genetically engineered yeast used in yeast surface display for protein and antibody screening. Derived from BJ5465, it contains genomic insertion of AGA1 under GAL promoter with a URA3 marker.
Catalog No. 50-238-3727 Supplier ATCC Supplier No. MYA4941
Saccharomyces cerevisiae strain EBY100 (ATCC MYA-4941™) is a genetically engineered yeast used in yeast surface display for protein and antibody screening. Derived from BJ5465, it contains genomic insertion of AGA1 under GAL promoter with a URA3 marker.