Learn More
ATCC Saccharomyces cerevisiae; purified DNA 5ug
Catalog No. 502381043
Encompass
Genomic DNA isolated from Saccharomyces cerevisiae BY4741 [ATCC 4040002]. This fungal strain is also available as ATCC Catalog No.: 201388™.
Catalog No. 50-238-1043 Supplier ATCC Supplier No. 201388D5
Add to Cart
Add to Cart
Genomic DNA isolated from Saccharomyces cerevisiae BY4741 [ATCC 4040002]. This fungal strain is also available as ATCC Catalog No.: 201388™.