Learn More
ATCC Saccharomyces cerevisiae; Strain: BCY123
Catalog No. 502384546
Encompass
Saccharomyces cerevisiae strain BCY123 is provided as an ampoule containing viable cells (yeast cells, spores, or agar cubes with mycelia) suspended in cryoprotectant. This strain can be used for protein production in yeast.
Catalog No. 50-238-4546 Supplier ATCC Supplier No. MYA5007
Saccharomyces cerevisiae strain BCY123 is provided as an ampoule containing viable cells (yeast cells, spores, or agar cubes with mycelia) suspended in cryoprotectant. This strain can be used for protein production in yeast.