Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saccharopine hydrochloride | 99.8% | 312.75 | C11H21ClN2O6 | 5 MG
SDP

Catalog No. 5000208507
Encompass_Preferred

Saccharopine hydrochloride is a lysine degradation intermediate that acts as a mitochondrial toxin and can disrupt mitochondrial homeostasis. It is supplied for research use in biochemical and cellular studies of lysine metabolism and mitochondrial function.

  • Lysine degradation intermediate and mitochondrial toxin.
  • Useful for research on lysine metabolism and mitochondrial function.
  • Molecular formula: C11H21ClN2O6.
  • Molecular weight: 312.75.
  • Purity: 99.8%.
  • Available in small milligram quantities for bench-scale experiments.

Catalog No. 50-002-08507 Supplier Medchemexpress LLC Supplier No. HYW040307B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saccharopine hydrochloride is a lysine degradation intermediate that acts as a mitochondrial toxin and can disrupt mitochondrial homeostasis. It is supplied for research use in biochemical and cellular studies of lysine metabolism and mitochondrial function.

  • Lysine degradation intermediate and mitochondrial toxin.
  • Useful for research on lysine metabolism and mitochondrial function.
  • Molecular formula: C11H21ClN2O6.
  • Molecular weight: 312.75.
  • Purity: 99.8%.
  • Available in small milligram quantities for bench-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.