Learn More
Medchemexpress LLC Saccharopine hydrochloride | 997-68-2 | MFCD34676410 | 99.8% | 312.75 g/mol | C11H21ClN2O6 | 50 MG
SDP
Catalog No. 5000208729
Encompass_Preferred
Saccharopine hydrochloride is the hydrochloride salt of L-saccharopine, a lysine degradation intermediate used as a research reagent to study lysine metabolism and mitochondrial function. It is a mitochondrial toxin that can impair development by disrupting mitochondrial homeostasis.
  • Used as a research reagent for studies of lysine degradation and mitochondrial biology.
  • High reported purity suitable for analytical and biological research.
  • Water-soluble; dissolves readily in water with ultrasonic assistance.
  • Stable when stored sealed at -20°C; in solution, long-term storage at -80°C is recommended.
  • Available in small mg package sizes for laboratory research use.
Catalog No. 50-002-08729 Supplier Medchemexpress LLC Supplier No. HYW040307B50MG
Saccharopine hydrochloride is the hydrochloride salt of L-saccharopine, a lysine degradation intermediate used as a research reagent to study lysine metabolism and mitochondrial function. It is a mitochondrial toxin that can impair development by disrupting mitochondrial homeostasis.
  • Used as a research reagent for studies of lysine degradation and mitochondrial biology.
  • High reported purity suitable for analytical and biological research.
  • Water-soluble; dissolves readily in water with ultrasonic assistance.
  • Stable when stored sealed at -20°C; in solution, long-term storage at -80°C is recommended.
  • Available in small mg package sizes for laboratory research use.