Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Saccharopine (hydrochloride) | MFCD34676410 | 99.8% | 312.75 g/mol | C11H21ClN2O6 | 100 MG
SDP

Catalog No. 5000209335
Encompass_Preferred

Saccharopine hydrochloride is a lysine degradation intermediate used in research as a mitochondrial toxin. It is supplied as a high-purity solid for in vitro research applications.

  • High purity, suitable for research use.
  • Molecular weight 312.75 g/mol.
  • Chemical formula C11H21ClN2O6.
  • Available in a 100 mg research pack.
  • Used as an intermediate in lysine metabolism studies and mitochondrial toxicity research.

Catalog No. 50-002-09335 Supplier Medchemexpress LLC Supplier No. HYW040307B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saccharopine hydrochloride is a lysine degradation intermediate used in research as a mitochondrial toxin. It is supplied as a high-purity solid for in vitro research applications.

  • High purity, suitable for research use.
  • Molecular weight 312.75 g/mol.
  • Chemical formula C11H21ClN2O6.
  • Available in a 100 mg research pack.
  • Used as an intermediate in lysine metabolism studies and mitochondrial toxicity research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.