Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples, Ricca Chemical
SDP

Catalog No. 66555 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6655-5 20 L Cubitainer
6655-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6655-5 Supplier Ricca Chemical Company Supplier No. 66555
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Physical Form Liquid
Chemical Name or Material Saccomanno Collection Fluid
Name Note Fixative and Preservative for Sputum Samples
CAS 7732-18-5, 64-17-5, 25322-68-3, 67-56-1, 3844-45-9
CAS Min % 53.0, 42.28, 2.3, 5.0E-4, 4.0E-4
CAS Max % 56.0, 44.01, 2.4, 5.0E-4, 4.0E-4
Packaging Cubitainer
Color Green
Quantity 20 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.