Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Saccomanno Cytology Collection Fluid
SDP

Catalog No. 21402153
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-402-153 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-153 Supplier StatLab Supplier No. SL2516
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

A ready-to-use fixative used for the collection of sputum

  • Lyses red blood cells
  • Mucoproteins remain suspended
  • Cells from samples are easily concentrated by centrifuging or filtering
  • Cells display excellent staining characteristics
  • 16 oz

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.