Learn More
INNOVATIVE BIOCHIPS Single Cell Isolation Chip: 1CellDish-60mm, 64 wells, Pack of 5
Catalog No. nc1637881
Encompass_Preferred
1CellDish is a handheld device with microfluidics aliquoting channels designed for single-cell isolation. The operation is by pipetting cell suspension into the center well and distributing cells to the satellite wells to form single-cell aliquots in less than 2 µL volumes. Features: 1) It provides rapid, convenient, and damage-free cell isolation. 2) It simplifies steps for harvesting target cells and genes. 3) It is compatible with all cell sizes smaller than 50 µm at either low or high cell density. 4) It uniformly distributes cell suspension into 64 outlet wells and retrieves single target cells by pipetting. Parameters: 1) Size of device: standard 60 mm Petri Dish format. 2) Number of wells: 64. 3) Well volume: 2.5 µL. 4) Single-cell yield: about 20 cells per device. Summary: 1CellDish isolates cells at a price below $5 per cell. It is used for single-cell lysis, PCR, sequencing, and multiomics.
Catalog No. NC1637881 Supplier INNOVATIVE BIOCHIPS Supplier No. H2SCD5PK
1CellDish is a handheld device with microfluidics aliquoting channels designed for single-cell isolation. The operation is by pipetting cell suspension into the center well and distributing cells to the satellite wells to form single-cell aliquots in less than 2 µL volumes. Features: 1) It provides rapid, convenient, and damage-free cell isolation. 2) It simplifies steps for harvesting target cells and genes. 3) It is compatible with all cell sizes smaller than 50 µm at either low or high cell density. 4) It uniformly distributes cell suspension into 64 outlet wells and retrieves single target cells by pipetting. Parameters: 1) Size of device: standard 60 mm Petri Dish format. 2) Number of wells: 64. 3) Well volume: 2.5 µL. 4) Single-cell yield: about 20 cells per device. Summary: 1CellDish isolates cells at a price below $5 per cell. It is used for single-cell lysis, PCR, sequencing, and multiomics.