Learn More
Mettler Toledo™ Sachet Holder, EasyPlace, Rotating
Catalog No. 01911935 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-935 EasyPlace
1 options
This item is not returnable. View return policy
Catalog No. 01-911-935 Supplier Mettler Toledo™ Supplier No. 30671573
Add to Cart
Add to Cart
This item is not returnable. View return policy
The sachet holder can hold up to three buffer sachets and prevent spillage. It can be rotated to place one buffer after the other in the calibration position.
  • Safe Sachet Handling
  • The rotating sachet holder sits securely on the EasyPlace baseplate. It holds up to three standard sachets, preventing spills
  • Straightforward Calibration
  • The placement on the EasyPlace makes sensor insertion simple. A simple turn of the holder moves the next standard into the measurement position
TRUSTED_SUSTAINABILITY
Specifications
Type Sachet Holder, EasyPlace Rotating
For Use With (Equipment) EasyPlace