Learn More
New England Biolabs, Inc. SacII – 10000 units
SDP
Catalog No. 50812057
Encompass
A restriction endonuclease that recognizes the sequence CC_GC^GG.
Catalog No. 50-812-057 Supplier New England Biolabs, Inc. Supplier No. R0157L
A restriction endonuclease that recognizes the sequence CC_GC^GG.