Learn More
TARGETMOL CHEMICALS INC SACLOFEN 5MG
SDP
Catalog No. 502576763
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Saclofen is a sulfonic analog of the inhibitory neurotransmitter (gamma)-aminobutyric acid (GABA) that acts as a competitive antagonist of the GABAB receptor (IC50 7.8 uM).1 This compound can be used to determine the functional roles for the GABAB receptor as a mediator of slow inhibitory postsynaptic potentials in the brain. purity: 99%
Catalog No. 50-257-6763 Supplier TARGETMOL CHEMICALS INC Supplier No. T44405MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Saclofen is a sulfonic analog of the inhibitory neurotransmitter (gamma)-aminobutyric acid (GABA) that acts as a competitive antagonist of the GABAB receptor (IC50 7.8 uM).1 This compound can be used to determine the functional roles for the GABAB receptor as a mediator of slow inhibitory postsynaptic potentials in the brain. purity: 99%