Learn More
Medchemexpress LLC Sacubitril-d4 | 1884269-07-1 | C24H25D4NO5 | 1 MG
SDP
Catalog No. 5000376618
Encompass_Preferred
Sacubitril-d4 is the deuterium-labeled version of Sacubitril, a potent Neprilysin (NEP) inhibitor (IC50 of 5 nM) and a component of the heart failure medicine LCZ696. This compound is valuable for research, serving as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Supports research in heart failure
Catalog No. 50-003-76618 Supplier Medchemexpress LLC Supplier No. HY15407S1MG
Sacubitril-d4 is the deuterium-labeled version of Sacubitril, a potent Neprilysin (NEP) inhibitor (IC50 of 5 nM) and a component of the heart failure medicine LCZ696. This compound is valuable for research, serving as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Supports research in heart failure