Learn More
Medchemexpress LLC Sacubitril-d4 hemicalcium salt | 1369773-39-6 | C24H25D4NO50.5Ca | 1 MG
SDP
Catalog No. 5000376622
Encompass_Preferred
Sacubitril-d4 hemicalcium salt is the deuterium labeled Sacubitril hemicalcium salt. Sacubitril hemicalcium salt is a potent NEP inhibitor with an IC50 of 5 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-76622 Supplier Medchemexpress LLC Supplier No. HY15407AS11MG
Sacubitril-d4 hemicalcium salt is the deuterium labeled Sacubitril hemicalcium salt. Sacubitril hemicalcium salt is a potent NEP inhibitor with an IC50 of 5 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.