Learn More
Medchemexpress LLC Sacubitril-d4 hemicalcium salt | 1369773-39-6 | C24H25D4NO50.5Ca | 5 MG
SDP
Catalog No. 5000376623
Encompass_Preferred
Sacubitril-d4 hemicalcium salt is the deuterium labeled version of Sacubitril hemicalcium salt. It is recognized as a potent NEP (Neprilysin) inhibitor, with an IC50 of 5 nM. Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development, and deuteration can influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-76623 Supplier Medchemexpress LLC Supplier No. HY15407AS15MG
Sacubitril-d4 hemicalcium salt is the deuterium labeled version of Sacubitril hemicalcium salt. It is recognized as a potent NEP (Neprilysin) inhibitor, with an IC50 of 5 nM. Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development, and deuteration can influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.