Learn More
Selleck Chemical LLC Sacubitril hemicalcium salt S4426-1g
SDP
Supplier:  Selleck Chemical LLC S44261G
Encompass_Preferred
Sacubitril (AHU-377) hemicalcium salt a component of the heart failure medicine LCZ696 is a potent inhibitor of neprilysin (neutral endopeptidase NEP)
Catalog No. 50-311-2195