Learn More
Selleck Chemical LLC Sacubitril hemicalcium salt S4426-25mg
SDP
Catalog No. 503110250
Encompass_Preferred
Sacubitril (AHU-377) hemicalcium salt a component of the heart failure medicine LCZ696 is a potent inhibitor of neprilysin (neutral endopeptidase NEP)
Catalog No. 50-311-0250 Supplier Selleck Chemical LLC Supplier No. S442625MG
Sacubitril (AHU-377) hemicalcium salt a component of the heart failure medicine LCZ696 is a potent inhibitor of neprilysin (neutral endopeptidase NEP)