Learn More
TARGETMOL CHEMICALS INC Sacubitril hemicalcium salt
SDP
Catalog No. 502878765
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Sacubitril hemicalcium salt (AHU377 calcium salt) is a potent NEP inhibitor with an IC50 of 5 nM. Sacubitril hemicalcium salt is a component of the heart failure medicine LCZ696. Sacubitril hemicalcium salt is a prodrug of LBQ657, which is an inhibitor of the zinc metallopeptidase neprilysin. Neprilysin degrades a variety of vasoactive peptides such as atrial and brain natriuretic peptide, bradykinin, adrenomedullin, and endothelin-1. Inhibition of neprilysin leads to an increased level of these peptides and, thus, antihypertensive effects. Formulations containing AHU377 in combination with the angiotensin II receptor antagonist valsartan are used to treat hypertension and heart failure. Purity 99.93%
Catalog No. 50-287-8765 Supplier TARGETMOL CHEMICALS INC Supplier No. T420050MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Sacubitril hemicalcium salt (AHU377 calcium salt) is a potent NEP inhibitor with an IC50 of 5 nM. Sacubitril hemicalcium salt is a component of the heart failure medicine LCZ696. Sacubitril hemicalcium salt is a prodrug of LBQ657, which is an inhibitor of the zinc metallopeptidase neprilysin. Neprilysin degrades a variety of vasoactive peptides such as atrial and brain natriuretic peptide, bradykinin, adrenomedullin, and endothelin-1. Inhibition of neprilysin leads to an increased level of these peptides and, thus, antihypertensive effects. Formulations containing AHU377 in combination with the angiotensin II receptor antagonist valsartan are used to treat hypertension and heart failure. Purity 99.93%