Learn More
Medchemexpress LLC Sacubitrilat | 149709-44-4 | 99.1% | 383.44 | 100 MG
SDP
Catalog No. 5000386987
Encompass_Preferred
Sacubitrilat (Desethyl Sacubitril) is an active neprilysin (NEP) inhibitor, designed for research use only. It is a single diastereomer that binds to the active site of NEP with high inhibitory potency.
  • Active neprilysin (NEP) inhibitor
  • Single diastereomer with specific stereocenters
  • High inhibitory potency (5 nM)
  • For research use only
Catalog No. 50-003-86987 Supplier Medchemexpress LLC Supplier No. HY17620100MG
Sacubitrilat (Desethyl Sacubitril) is an active neprilysin (NEP) inhibitor, designed for research use only. It is a single diastereomer that binds to the active site of NEP with high inhibitory potency.
  • Active neprilysin (NEP) inhibitor
  • Single diastereomer with specific stereocenters
  • High inhibitory potency (5 nM)
  • For research use only