Learn More
Medchemexpress LLC Sacubitrilat-d4 | 149709-44-4 | 99.3% | 387.46 | C22H21D4NO5 | 10 MG
SDP
Catalog No. 5000298714
Encompass_Preferred
Sacubitrilat-d4 is the deuterium-labeled form of sacubitrilat (desethyl sacubitril), provided as a stable-isotope reference standard for bioanalytical and drug-development studies. It is intended for research use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS assays.
  • Deuterium-labeled isotopologue for use as an internal standard.
  • High chemical purity: 99.3%.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Supplied as a 10 mg powder for analytical applications.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).
Catalog No. 50-002-98714 Supplier Medchemexpress LLC Supplier No. HY17620S10MG
Sacubitrilat-d4 is the deuterium-labeled form of sacubitrilat (desethyl sacubitril), provided as a stable-isotope reference standard for bioanalytical and drug-development studies. It is intended for research use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS assays.
  • Deuterium-labeled isotopologue for use as an internal standard.
  • High chemical purity: 99.3%.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Supplied as a 10 mg powder for analytical applications.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).