Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sacubitrilat-d4 | 99.3% | 387.46 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY17620S1MG

Encompass_Preferred

Sacubitrilat-d4 is the deuterium labeled Sacubitrilat. Sacubitrilat (Desethyl Sacubitril) is an active neprilysin (NEP) inhibitor. This compound is for research use only and not sold to patients.

  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution: stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Purity: 99.28%
  • Appearance: solid, white to off-white.
  • Solubility: soluble in DMSO at 100 mg/mL (258.09 mM).
  • Storage: powder at -20°C for 3 years or 4°C for 2 years. In solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-87433


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.