Learn More
Chemglass Life Sciences Saddles, Berl, 200cc, Porcelain, Distilling Column, Approx. 250 Pieces/Package
SDP
Catalog No. 50871939
Encompass_Preferred
Saddles, Berl, 200cc, Porcelain, Distilling Column, Approx. 250 Pieces/Package. Porcelain berl saddles having an O.D. of 8mm. Ideal for vacuum distillation, absorption and extraction. Saddles provide a highly useable surface area, lower gas flow resistance and excellent internal distribution. Available in packages of 200cc and 1000cc boxes.
Catalog No. 50-871-939 Supplier Chemglass Life Sciences Supplier No. CG128401
Saddles, Berl, 200cc, Porcelain, Distilling Column, Approx. 250 Pieces/Package. Porcelain berl saddles having an O.D. of 8mm. Ideal for vacuum distillation, absorption and extraction. Saddles provide a highly useable surface area, lower gas flow resistance and excellent internal distribution. Available in packages of 200cc and 1000cc boxes.