Learn More
Aviva Systems Biology Corp SAE1 Antibody - N-terminal region (ARP52084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SAE1. Alternate names: AOS1, FLJ3091, HSPC140, SUA1, UBLE1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53874