Learn More
Texas Medical Industries SAF-EVAC 1000 PK
SDP
Catalog No. NC0455422
Encompass
SAF-EVAC™ this patented device prevents the splashing of blood from uncapping sampling tubes. For use with Point of Care diagnostic systems including the i-STAT™ and laboratory hematology slides. SAF-EVAC allows laboratory personnel to dispense whole anti-coagulated blood from the tube one drop at a time. The blood smear may now be prepared without removal of the stopper. SAF-EVAC helps laboratories meet OSHA regulations criteria dated December 6, 1991, 29 CFR part 1910.1030, Occupational Exposure to Blood borne Pathogens: Final Rule. Excerpt from rule: "All procedures involving blood or other potentially infectious materials shall be performed in such a manner as to minimize splashing, spraying, and generation of droplets of these substan
Catalog No. NC0455422 Supplier Texas Medical Industries Supplier No. 5004
SAF-EVAC™ this patented device prevents the splashing of blood from uncapping sampling tubes. For use with Point of Care diagnostic systems including the i-STAT™ and laboratory hematology slides. SAF-EVAC allows laboratory personnel to dispense whole anti-coagulated blood from the tube one drop at a time. The blood smear may now be prepared without removal of the stopper. SAF-EVAC helps laboratories meet OSHA regulations criteria dated December 6, 1991, 29 CFR part 1910.1030, Occupational Exposure to Blood borne Pathogens: Final Rule. Excerpt from rule: "All procedures involving blood or other potentially infectious materials shall be performed in such a manner as to minimize splashing, spraying, and generation of droplets of these substan